Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cystine, FCC, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18603745 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-745 100 g Poly Bottle
18-603-744 25 g Poly Bottle
18-603-746 1 kg Poly Pail
18-603-747 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-745 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1476100GM
Add to Cart
Add to Cart

C1476, 56-89-3, C6H12N2O4S2

Spectrum™ Chemical L-Cystine, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56-89-3
Molecular Formula C6H12N2O4S2
Molecular Weight (g/mol) 240.29
InChI Key LEVWYRKDKASIDU-IMJSIDKUSA-N
IUPAC Name (2R)-2-amino-3-{[(2R)-2-amino-2-carboxyethyl]disulfanyl}propanoic acid
SMILES N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O

Specifications

CAS Max % 100%
Quantity 100 g
Percent Purity 98.5 to 101.5%
Grade FCC
Ignition Residue 0.1%
Loss on Drying 0.2%
Packaging Poly Bottle
Specific Rotation -215° to -225°
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.