Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Diacetyltartaric acid AM resin, 1% DVB, 0.9-1.1mmol/g, 100-200 mesh, Thermo Scientific™

Catalog No. ac361150050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS -01-1
MDL Number MFCD03457311
Synonym L-Diacetyltartaric acid Aminomethylpolystyrene

Specifications

Color Beige to White or Brown
Physical Form Beads
Loading 0.9 to 1.1mmol/g
Mesh Size 100 to 200 mesh
Packaging Glass bottle
Quantity 5 g
Chemical Name or Material L-Diacetyltartaric acid AM resin, 1% DVB, 0.9-1.1mmol/g
Recommended Storage Refrigerator +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.