Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(-)-epinephrine-(+)-bitartrate, MP Biomedicals™

Catalog No. ICN19516625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19516625 25 g
ICN19516605 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19516625 Supplier MP Biomedicals, Inc Supplier No. 0219516625

An adrenergic receptor agonist

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-42-3
Molecular Formula C13H19NO9
Molecular Weight (g/mol) 333.29
MDL Number MFCD00035077
InChI Key YLXIPWWIOISBDD-NDAAPVSOSA-N
Synonym L-Adrenalin Bitartrate, Adrenaline Acid Tartrate
PubChem CID 124080922
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid; 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
SMILES O[C@H]([C@@H](O)C(O)=O)C(O)=O.CNC[C@H](O)C1=CC=C(O)C(O)=C1

Specifications

Color White
Quantity 25 g
Formula Weight 333.29
Physical Form Powder
Chemical Name or Material L-(-)-epinephrine-(+)-bitartrate
EINECSNumber 200-097-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.