Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Osmium tetroxide, 99.9+%, (trace metal basis)
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Oxidation reagent
Specifications
Specifications
| Boiling Point | 130.0°C |
| Melting Point | 40.0°C to 42.0°C |
| Color | Colorless to Yellow |
| Physical Form | Crystals |
| Quantity | 5 g |
| Assay Percent Range | 99.9+% |
| Linear Formula | OsO4 |
| Fieser | 01,759; 02,301; 04,361; 05,141; 06,424; 10,290; 12,358; 13,186; 14,235; 15,240; 16,249; 17,236 |
| Merck Index | 15, 6990 |
| Solubility Information | Passes test |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Fatal in contact with skin. Causes severe skin burns and eye damage. Fatal if inhaled. Fatal if swallowed. May cause allergy or asthma symptoms or breathing difficulties if inhaled. |
| Precautionary Statement | GHS P Statement Rinse mouth. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/f |
| EINECSNumber | 244-058-7 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction