Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(-)-Fenchone, 98+%

Catalog No. p-133338
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle

CAS: 7787-20-4 | C10H16O | 152.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-20-4
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00151104
InChI Key LHXDLQBQYFFVNW-GMSGAONNSA-N
Synonym l-fenchone, 1r-fenchone, 1r,4r-2,2,4-trimethylbicyclo 2.2.1 heptan-3-one, 2r,5r-2,6,6-trimethyl-2,5-methanocyclohexane-1-one
PubChem CID 3034206
IUPAC Name (1R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C2CCC(C2)(C1=O)C)C

Specifications

Melting Point 5°C
Color Colorless to Yellow
Density 0.9400g/mL
Boiling Point 192°C to 194°C
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4610 to 1.463
Quantity 50 g
Fieser 08,228
Specific Gravity 0.94
Specific Rotation Condition −50.50° (24°C neat)
Specific Rotation -50.5
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether
Formula Weight 152.24
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material L(-)-Fenchone, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 232-107-5
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.