Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ L-Ficolin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120478
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120478 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120478 Supplier Invitrogen™ Supplier No. PA5120478
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: mosue spleen, mouse lung, rat spleen. Immunogen sequence: SAGDSLTFHN NQSFSTKDQD NDLNTGNCAV MFQGAWWYKN CHVSNLNGRY LRGTHGSFAN G.

L-Ficolin (Ficolin-2) is an innate immunity pattern recognition molecule found in human serum. L-Ficolin binds to distinct pathogen-associated molecular patterns (PAMP), such as carbohydrates (GlcNAc), lipoteichoic acid and acetylated groups thereby facilitating phagocytosis and activation of complement through the lectin route using the same serine proteases as mannose-binding lectin (MBL) named MASP ((MBL)-associated serine protease).
TRUSTED_SUSTAINABILITY

Specifications

Antigen L-Ficolin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FCN2
Gene Accession No. O70497, Q15485
Gene Alias 37 kDa elastin-binding protein; Collagen/fibrinogen domain-containing protein 2; EBP-37; FCN2; Fcnb; FCNL; ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin); ficolin 2; ficolin B; ficolin-2; Ficolin-B; ficolin-beta; Hucolin; L-ficolin; P35; RP11-263F14.2; Serum lectin p35
Gene Symbols FCN2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 230-290 of human FCN2 (NP_004099.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14134, 2220
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.