Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(-)-Glucose, anhydrous, 98%, Thermo Scientific Chemicals

Catalog No. aaa1749603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1749603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1749603 Supplier Thermo Scientific Chemicals Supplier No. A1749603
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 921-60-8 | Molecular Formula: C6H12O6 | Molecular Weight: 180.16 g/mol

L-(-)-Glucose is an enantiomer of the more common D-glucose a naturally occurring carbohydrate used in an abundance of cellular processes. L-Glucose is a synthetic sugar used in the formation of L-Glucose Pen taacetate, a potential therapeutic agent regarding type II diabetes. In addition, L-glucose can be used as a colon cleansing agent for before a colonoscopy procedure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(-)-Glucose is an enantiomer of the more common D-glucose a naturally occurring carbohydrate used in an abundance of cellular processes. L-Glucose is a synthetic sugar used in the formation of L-Glucose Pen taacetate, a potential therapeutic agent regarding type II diabetes. In addition, L-glucose can be used as a colon cleansing agent for before a colonoscopy procedure.

Solubility
Soluble in water, alcohol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 921-60-8
Molecular Formula C6H12O6
MDL Number MFCD00148913
PubChem CID 10954115
ChEBI CHEBI:37626

Specifications

Melting Point 148°C to 157°C
Quantity 1 g
Beilstein 1724626
Synonym l-glucose, l---glucose, levoglucose, l--glucose, 2s,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, aldehydo-l-glucose, aldehydo-l-gluco-hexose, d-glucose-13c6,d7, levoglucose usan, glucose, l-isomer
Solubility Information Soluble in water,alcohol.
InChI Key GZCGUPFRVQAUEE-VANKVMQKSA-N
Optical Rotation −53° (c=10 in Water, 24h)
SMILES C(C(C(C(C(C=O)O)O)O)O)O
IUPAC Name (2S,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
Molecular Weight (g/mol) 180.156
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material L-(-)-Glucose, anhydrous
Show More Show Less
Hazard Category H500
EINECSNumber 213-068-3
RTECSNumber LZ6610000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.