Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Glutamic acid 1-methyl ester, 98%

Click to view available options
Quantity:
1g
5g
6384-08-3
C8H12N2
136.20
MFCD00171677
BWAPJIHJXDYDPW-UHFFFAOYSA-N
2,5-dimethyl-1,4-benzenediamine,4-amino-2,5-dimethylaniline,2,5-dimethyl-1,4-phenylenediamine,2,5-dimethyl-p-phenylenediamine,2,5-diamino-p-xylene,2,5-dimethylphenylene-1,4-diamine,2,5-dimethyl-1,4-benzoquinonediamine,ccris 8118,1,4-dimethylphenylene-2,5-diamine,unii-re400vn2b4
160824
CHEBI:53613
2,5-dimethylbenzene-1,4-diamine
CC1=CC(=C(C=C1N)C)N
6384-08-3
C8H12N2
136.20
MFCD00171677
BWAPJIHJXDYDPW-UHFFFAOYSA-N
2,5-dimethyl-1,4-benzenediamine,4-amino-2,5-dimethylaniline,2,5-dimethyl-1,4-phenylenediamine,2,5-dimethyl-p-phenylenediamine,2,5-diamino-p-xylene,2,5-dimethylphenylene-1,4-diamine,2,5-dimethyl-1,4-benzoquinonediamine,ccris 8118,1,4-dimethylphenylene-2,5-diamine,unii-re400vn2b4
160824
CHEBI:53613
2,5-dimethylbenzene-1,4-diamine
CC1=CC(=C(C=C1N)C)N
Solubility
Soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Beilstein 1707273
Solubility Information Soluble in water.
Formula Weight 161.16
Physical Form Powder
Percent Purity 98%
Color White
Melting Point 145°C (decomposition)
Quantity 1g
Chemical Name or Material L-Glutamic acid 1-methyl ester

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold