Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals L-Glutamic acid, 99%

Catalog No. AAA150310B
Change view
Click to view available options
Quantity:
250 g
500 g
1000 g
5000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA150310B 1000 g
AAA1503130 250 g
AAA1503136 500 g
AAA150310I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA150310B Supplier Thermo Scientific Chemicals Supplier No. A150310B
Add to Cart

CAS: 56-86-0 | C5H9NO4 | 147.13 g/mol

It is used as a chiral building-block in the synthesis of the (S)-isomer of -butyrolactone-4-carboxylic acid.  L-Glutamic acid is widely used in fields of medicine, food processing, industry, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a chiral building-block in the synthesis of the (S)-isomer of -butyrolactone-4-carboxylic acid.  L-Glutamic acid is widely used in fields of medicine, food processing, industry, etc.

Solubility
It is very slightly soluble in cold water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-86-0
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002634
InChI Key WHUUTDBJXJRKMK-UHFFFAOYNA-N
Synonym l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
PubChem CID 33032
ChEBI CHEBI:16015
IUPAC Name (2S)-2-aminopentanedioic acid
SMILES NC(CCC(O)=O)C(O)=O

Specifications

Melting Point 189°C to 192°C (decomposition)
Assay Percent Range ≥99%
Beilstein 1723801
Merck Index 14,4469
Solubility Information It is very slightly soluble in cold water.
Density 1.525
Quantity 1000 g
Formula Weight 147.13
Optical Rotation +31° (c=5 in 5N HCl)
Percent Purity ≥99%
Chemical Name or Material L-Glutamic acid
Show More Show Less
Hazard Category H500
EINECSNumber 200-293-7
RTECSNumber LZ9700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.