Learn More
L-Glutamic Acid Monopotassium Salt, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Poly Bottle |
| Quantity | 500 g |
| Grade | Technical |
| Chemical Name or Material | L-Glutamic Acid Monopotassium Salt |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.