Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva L-Glutamine
Click to view available options
Quantity:
100 mL
500 mL
Description
- Format: Liquid

Specifications
Specifications
Content And Storage | Store frozen |
Description | L-Glutamine, 200mM Solution (29.2mg/mL-Glutamine in 0.85% NaCl), 100mL |
Concentration | 29.2 mg/mL |
Form | Liquid |
Sterilization Method | 0.1 μm Sterile-filtered |
Quantity | 100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction