Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Glutamine, ≥98.5%, Ultrapure
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Packaging:
Plastic Bottle
Specifications
Specifications
| Packaging | Plastic Bottle |
| Linear Formula | H2NCOCH2CH2CH(NH2)COOH |
| Beilstein | 1723797 |
| Quantity | 250 g |
| Formula Weight | 146.14 |
| Percent Purity | ≥98.5% |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | L-Glutamine |
Safety and Handling
| Hazard Category | H315-H319-H320-H335-H336 |
| Precautionary Statement | P261-P264b-P271-P280g-P280i-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 200-292-1 |
| RTECSNumber | MA2275100 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction