Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamine, ≥98.5%, Ultrapure

Catalog No. p-7082818
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Packaging:
Plastic Bottle

CAS: 56-85-9 | C5H10N2O3 | 146.146 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.146
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
IUPAC Name (2 S)-2,5-diamino-5-oxopentanoic acid
SMILES C(CC(=O)N)C(C(=O)O)N

Specifications

Packaging Plastic Bottle
Linear Formula H2NCOCH2CH2CH(NH2)COOH
Beilstein 1723797
Quantity 100 g
Formula Weight 146.14
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material L-Glutamine
Hazard Category H315-H319-H320-H335-H336
Precautionary Statement P261-P264b-P271-P280g-P280i-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-292-1
RTECSNumber MA2275100
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.