Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ L-Glutamine, ≥98.5%, Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
100g
250g
500g
1kg
Packaging:
Plastic bottle
56-85-9
C5H10N2O3
146.146
MFCD00008044
ZDXPYRJPNDTMRX-VKHMYHEASA-N
l-glutamine,glutamine,levoglutamide,l-+-glutamine,glutamic acid amide,h-gln-oh,stimulina,cebrogen,glumin,levoglutamid
5961
CHEBI:18050
(2S)-2,5-diamino-5-oxopentanoic acid
C(CC(=O)N)C(C(=O)O)N
56-85-9
C5H10N2O3
146.146
MFCD00008044
ZDXPYRJPNDTMRX-VKHMYHEASA-N
l-glutamine,glutamine,levoglutamide,l-+-glutamine,glutamic acid amide,h-gln-oh,stimulina,cebrogen,glumin,levoglutamid
5961
CHEBI:18050
(2S)-2,5-diamino-5-oxopentanoic acid
C(CC(=O)N)C(C(=O)O)N
Linear Formula H2NCOCH2CH2CH(NH2)COOH
Packaging Plastic bottle
Formula Weight 146.14
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Quantity 1kg
Chemical Name or Material L-Glutamine