Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tert-Butyllithium, 1.9M solution in pentane, AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Yellow |
| Density | 0.6900g/mL |
| Boiling Point | 36.0°C to 38.0°C |
| Flash Point | −56°C |
| Packaging | AcroSeal™ Glass bottle |
| Linear Formula | (CH3)3CLi |
| Quantity | 800 mL |
| Specific Gravity | 0.69 |
| IUPAC Name | lithium(1+) 2-methylpropan-2-ide |
| Formula Weight | 64.04 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic to aquatic life with long lasting effects. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes severe skin burns and eye damage. Extremely flammable liquid and vapor |
| Precautionary Statement | GHS P Statement IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. Keep away from any possible contact with wa |
| EINECSNumber | 209-831-5 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction