Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-Glutamine (Laboratory), Fisher Chemical

Catalog No. o2956100 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
IUPAC Name (2S)-2-amino-4-carbamoylbutanoic acid
SMILES N[C@@H](CCC(N)=O)C(O)=O

Specifications

Melting Point 185°C
pH 4.0 to 6.0
Color White
Identification Pass Test
Packaging Glass Bottle
Linear Formula HO2CCH(NH2)CH2CH2CONH2
Alpha Vector Glutamine
Quantity 100 g
Formula Weight 146.15
Grade Laboratory
Physical Form Solid
Chemical Name or Material L-(+)-Glutamine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.