Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(-)-Glutathione, oxidized, 98%

Catalog No. p-138555
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 27025-41-8 | C20H32N6O12S2 | 612.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27025-41-8
Molecular Formula C20H32N6O12S2
Molecular Weight (g/mol) 612.63
MDL Number MFCD00150701
InChI Key YPZRWBKMTBYPTK-IOKZUGQQNA-N
Synonym oxiglutatione, glutathione disulfide, gssg, oxidized glutathione, glutathione disulphide, glutathione-ssg, l-glutathione oxidized, oxigluthione, glutathone disulfide, glutathione oxidized
PubChem CID 65359
ChEBI CHEBI:17858
IUPAC Name 2-amino-4-[(2-{[2-(4-amino-4-carboxybutanamido)-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl)carbamoyl]butanoic acid
SMILES N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O

Specifications

Melting Point 178.0°C
Color White
Ignition Residue 0.5% max. (800°C)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Beilstein 04, IV, 3168
Merck Index 15, 4511
Specific Rotation Condition −99.00 (20.00°C c=4,water)
Specific Rotation − 99.00
Solubility Information Solubility in water: soluble.
Formula Weight 612.64
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L(-)-Glutathione, oxidized, 98%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 248-170-7
RTECSNumber MC0556500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.