Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutathione, Reduced, 98.0-101.0%, Spectrum™ Chemical
SDP

Catalog No. 18611275 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
250 g
500 g
1 kg
5 kg
10 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-275 100 g Amber Glass Bottle
18-611-272 1 g Amber Glass Bottle
18-611-278 5 g Amber Glass Bottle
18-611-276 25 g Amber Glass Bottle
18-611-277 250 g Amber Glass Bottle
18-611-280 500 g Amber Glass Bottle
18-611-273 1 kg Amber Glass Bottle
18-611-279 5 kg Poly Pail
18-611-274 10 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18-611-275 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. GL146100GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 70-18-8
Molecular Formula C10H17N3O6S
Molecular Weight (g/mol) 307.32
InChI Key RWSXRVCMGQZWBV-UHFFFAOYNA-N
IUPAC Name 2-amino-4-({1-[(carboxymethyl)carbamoyl]-2-sulfanylethyl}carbamoyl)butanoic acid
SMILES NC(CCC(=O)NC(CS)C(=O)NCC(O)=O)C(O)=O

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 100 g
Specific Rotation -17.5° to -15.5°
Percent Purity 98 to 101%
Grade Reagent

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.