Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutathione, Reduced, 98.0-101.0%, Spectrum™ Chemical
SDP

Catalog No. 18611277
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
250 g
500 g
1 kg
5 kg
10 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

Chemical Identifiers

CAS 70-18-8
Molecular Formula C10H17N3O6S
Molecular Weight (g/mol) 307.32
InChI Key RWSXRVCMGQZWBV-UHFFFAOYNA-N
IUPAC Name 2-amino-4-({1-[(carboxymethyl)carbamoyl]-2-sulfanylethyl}carbamoyl)butanoic acid
SMILES NC(CCC(=O)NC(CS)C(=O)NCC(O)=O)C(O)=O

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 250 g
Specific Rotation -17.5° to -15.5°
Percent Purity 98 to 101%
Grade Reagent

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.