Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Histidinamide dihydrochloride, 95%, Thermo Scientific™

Catalog No. p-7056562
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71666-95-0
Molecular Formula C6H12Cl2N4O
Molecular Weight (g/mol) 227.089
MDL Number MFCD00058559
InChI Key CRBYFJCXMZNLTO-XRIGFGBMSA-N
Synonym h-his-nh2.2hcl, l-histidinamide dihydrochloride, h-his-nh2 2hcl, h-his-nh 2 hcl, c6h10n4o.2hcl, h-his-nh2 . 2 hcl, l-histidine amide dihydrochloride, l-histidinamide-hydrogen chloride 1/2, h-his-nh2 inverted exclamation mark currency 2 hcl, 2s-2-amino-3-1h-imidazol-4-yl propanamide dihydrochloride
PubChem CID 56777274
IUPAC Name (2S)-2-amino-3-(1H-imidazol-5-yl)propanamide;dihydrochloride
SMILES C1=C(NC=N1)CC(C(=O)N)N.Cl.Cl

Specifications

Beilstein 3720223
Quantity 250 mg
Formula Weight 227.09
Percent Purity 95%
Physical Form Solid
Chemical Name or Material L-Histidinamide dihydrochloride
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.