Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Histidine 99.0+%, TCI America™
SDP

Catalog No. H014925G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Melting Point 281°C
CAS 71-00-1
Color White
Molecular Formula C6H9N3O2
MDL Number MFCD00064315
Synonym l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
InChI Key HNDVDQJCIGZPNO-UHFFFAOYNA-N
SMILES NC(CC1=CN=CN1)C(O)=O
IUPAC Name 2-amino-3-(1H-imidazol-5-yl)propanoic acid
Molecular Weight (g/mol) 155.16
PubChem CID 6274
ChEBI CHEBI:15971
Formula Weight 155.16
Percent Purity ≥99.0% (HPLC,T)
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material L-Histidine
Show More Show Less
EINECSNumber (9)-1607
RTECSNumber MS3070000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.