Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Homoserine, 99%

Catalog No. aal12427md
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 672-15-1 | C4H9NO3 | 119.12 g/mol

L-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.

Solubility
Soluble in water (1100 mg/ml at 30°C), and methanol (Sparingly). Insoluble in ethanol.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 672-15-1
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00063090
InChI Key UKAUYVFTDYCKQA-VKHMYHEASA-N
Synonym l-homoserine, homoserine, s-2-amino-4-hydroxybutanoic acid, 2s-2-amino-4-hydroxybutanoic acid, h-hoser-oh, s-2-amino-4-hydroxybutyric acid, homoserine van, 2-amino-4-hydroxybutyric acid, s-homoserine, unii-6ka95x0ivo
PubChem CID 12647
ChEBI CHEBI:15699
IUPAC Name (2S)-2-amino-4-hydroxybutanoic acid
SMILES C(CO)C(C(=O)O)N

Specifications

Melting Point ∽203°C (decomposition)
Beilstein 1721681
Merck Index 14,4739
Solubility Information Soluble in water (1100mg/ml at 30°C),and methanol (Sparingly). Insoluble in ethanol.
Quantity 250 mg
Formula Weight 119.12
Optical Rotation −9° (c=2 in Water)
Percent Purity 99%
Chemical Name or Material L-Homoserine
EINECSNumber 211-590-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.