Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Isoleucine ethyl ester hydrochloride, 98%

Click to view available options
Quantity:
1g
5g
25g
56782-52-6
C8H18ClNO2
195.687
MFCD09953729
QQGRWNMNWONMOO-LEUCUCNGSA-N
l-isoleucine ethyl ester hydrochloride,h-ile-oet.hcl,ethyl l-isoleucinate hydrochloride,l-isoleucine ethyl ester hcl,2s,3s-ethyl 2-amino-3-methylpentanoate hydrochloride,isoleucine, ethyl ester, hydrochloride,ethyl 2s,3s-2-amino-3-methylpentanoate hydrochloride,l-isoleucine ethylester hcl,ksc497o6p,l-isoleucineethylesterhydrochloride
6453390
ethyl (2S,3S)-2-amino-3-methylpentanoate;hydrochloride
CCC(C)C(C(=O)OCC)N.Cl
56782-52-6
C8H18ClNO2
195.687
MFCD09953729
QQGRWNMNWONMOO-LEUCUCNGSA-N
l-isoleucine ethyl ester hydrochloride,h-ile-oet.hcl,ethyl l-isoleucinate hydrochloride,l-isoleucine ethyl ester hcl,2s,3s-ethyl 2-amino-3-methylpentanoate hydrochloride,isoleucine, ethyl ester, hydrochloride,ethyl 2s,3s-2-amino-3-methylpentanoate hydrochloride,l-isoleucine ethylester hcl,ksc497o6p,l-isoleucineethylesterhydrochloride
6453390
ethyl (2S,3S)-2-amino-3-methylpentanoate;hydrochloride
CCC(C)C(C(=O)OCC)N.Cl
Beilstein 4710859
Formula Weight 195.69
Physical Form Powder
Percent Purity 98%
Color White
Quantity 25g
Chemical Name or Material L-Isoleucine ethyl ester hydrochloride