Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-Lactic acid lithium salt, 99%, MP Biomedicals™

Catalog No. icn10213410
Click to view available options
Quantity:
10 g
100 g

Chemical Identifiers

CAS 867-55-0
Molecular Formula C3H5LiO3
Molecular Weight (g/mol) 96.01
InChI Key GKQWYZBANWAFMQ-UHFFFAOYSA-M
Synonym lithium lactate, lithium 2-hydroxypropanoate, lactic acid, lithium salt, dl-lactic acid lithium salt, propanoic acid, 2-hydroxy-, monolithium salt, lithium 1+ ion lactate, l-+-lactic acid lithium salt, lithotab lactate, acmc-20akmj, lithium s-lactate
PubChem CID 23673452
IUPAC Name lithium;2-hydroxypropanoate
SMILES [Li+].CC(C(=O)[O-])O

Specifications

Color White
Formula Weight 96.1
Percent Purity 99%
Quantity 10 g
Physical Form Powder
Chemical Name or Material L-(+)-Lactic acid lithium salt
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.