Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(-)-Lactic acid, lithium salt, 99%, pure

Catalog No. p-152927
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle

CAS: 27848-80-2 | C3H5O3Li | 96.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27848-80-2
Molecular Formula C3H5O3Li
Molecular Weight (g/mol) 96.01
MDL Number MFCD00065512
InChI Key GKQWYZBANWAFMQ-DKWTVANSSA-M
Synonym lithium s-2-hydroxypropanoate, l-lactic acid lithium salt, lithium l-lactate, s-2-hydroxypropionic acid lithium salt, sarcolactic acid lithium salt, lithium 1+ l-+-lactate, lithiuml-lactate, lithotab l-lactate, lactic acid lithium salt, c3h5o3.li
PubChem CID 23687877
IUPAC Name lithium;(2S)-2-hydroxypropanoate
SMILES [Li+].CC(C(=O)[O-])O

Specifications

Melting Point >75.0°C
Color White
Boiling Point >200.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3CH(OH)CO2Li
Quantity 5 g
Specific Rotation Condition −14.5° (20°C c=1,H2O)
Specific Rotation − 14.50
Solubility Information Solubility in water: 50g/100mL
Formula Weight 96.01
Percent Purity 99%
Grade Pure
Physical Form Fine Crystalline Powder
Chemical Name or Material L(-)-Lactic acid, lithium salt, 99%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 248-692-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.