Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Lactide, 98%, Thermo Scientific Chemicals

Catalog No. ac449210050
Click to view available options
Quantity:
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4511-42-6
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.13
MDL Number MFCD00070594
InChI Key JJTUDXZGHPGLLC-UHFFFAOYNA-N
Synonym l-lactide, 3s,6s-3,6-dimethyl-1,4-dioxane-2,5-dione, l---lactide, lactide, l, l-dilactide, 3s-cis-3,6-dimethyl-1,4-dioxane-2,5-dione, ll-lactide, unii-ij13to4no1, 1,4-dioxane-2,5-dione, 3,6-dimethyl-, 3s,6s, ij13to4no1
PubChem CID 107983
IUPAC Name (3S,6S)-3,6-dimethyl-1,4-dioxane-2,5-dione
SMILES CC1OC(=O)C(C)OC1=O

Specifications

Melting Point 95°C to 98°C
Boiling Point 255°C
Quantity 5 g
Packaging Glass bottle
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 144.13
Percent Purity 98%
Chemical Name or Material L-Lactide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 224-832-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.