Learn More
L-Leucine, 98.5 to 101.5% (Dry Basis), Ultrapure
Specifications
Specifications
| Melting Point | >300°C |
| Packaging | Plastic Bottle |
| Beilstein | 1721722 |
| Density | 1.293 |
| Quantity | 1 kg |
| Formula Weight | 131.17 |
| Percent Purity | 98.5 to 101.5% |
| Specific Gravity | 1.293 |
| Physical Form | Crystalline Powder |
| Assay | Dry basis |
| Show More |
Safety and Handling
| EINECSNumber | 200-522-0 |
| RTECSNumber | OH2850000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.