Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ L-Lysine-2HCl, 13C6, 15N2 for SILAC, 50 mg PROMO

Catalog No. PI88209PM
Click to view available options
:
Thermo Scientific™ L-Lysine-2HCl, 13C6, 15N2 for SILAC

Study protein expression by mass spectrometry with mass variants of lysine, arginine and other amino acids to perform stable isotope labeling with amino acids in cell culture.

SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of heavy 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.

Highlights:

  • Efficient – 100% label incorporation into proteins of living cells

  • Flexible – different isotopes of heavy and/or light amino acids for arginine, lysine, leucine and proline enable the quantitation of peptides derived from MS-grade proteases

  • Multiplex capabilities – several alternative isotopes of arginine and lysine are available that allow the analysis of multiple treatment conditions in each experiment

  • High-quality supplements – heavy amino acids with >99% isotope purity

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Amino Acids
Content And Storage Store at room temperature.
Label or Dye Mass Isotope of Amino Acid
Shipping Condition Room Temperature
Starting Material Cells
Final Product Type Protein
Detection Method Mass Spectrometry
Workflow Step Protein Labeling
Quantity 50 mg
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.