Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Lysine ethyl ester dihydrochloride, 99%

Catalog No. AAA1682414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1682414 25 g
AAA1682422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1682414 Supplier Thermo Scientific Chemicals Supplier No. A1682414
Add to Cart

CAS: 3844-53-9 | C8H20Cl2N2O2 | 247.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3844-53-9
Molecular Formula C8H20Cl2N2O2
Molecular Weight (g/mol) 247.16
MDL Number MFCD00039068
InChI Key DZIYAIZKJOHVQC-KLXURFKVSA-N
Synonym l-lysine ethyl ester dihydrochloride, h-lys-oet.2hcl, ethyl l-lysinate dihydrochloride, ethyl 2s-2,6-diaminohexanoate dihydrochloride, ethyl l-lysinate hcl, s-ethyl 2,6-diaminohexanoate dihydrochloride, l-lysine, ethyl ester, dihydrochloride, h-lys-oet·2hcl, h-lys-oet inverted exclamation mark currency2hcl, lys-oet2hcl
PubChem CID 107468
IUPAC Name ethyl (2S)-2,6-diaminohexanoate;dihydrochloride
SMILES Cl.Cl.CCOC(=O)[C@@H](N)CCCCN

Specifications

Melting Point ∼145°C (decomposition)
Beilstein 3697099
Quantity 25 g
Formula Weight 247.17
Percent Purity 99%
Sensitivity Hygroscopic
Chemical Name or Material L-Lysine ethyl ester dihydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-340-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.