Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-Lysine, Hydrate, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18611168
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 39665-12-8

Specifications

Ammonium 200 ppm
CAS Max % 1
pH 9.0 to 10.5
Chloride 300 ppm
Heavy Metals 10 ppm
Iron (Fe) 10 ppm
Other Amino Acids 0.005
Packaging Amber Glass Bottle
Specific Rotation +25.5° to+27.0°
Sulfate 200 ppm
Water 10.0 - 11.5%
Quantity 1 kg
Percent Purity 98.5 to 101.5%
Grade Reagent
Chemical Name or Material L-(+)-Lysine Hydrate
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.