Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-m-Tyrosine, 97+%

Catalog No. p-7029728
Click to view available options
Quantity:
250 mg
1 g

CAS: 587-33-7 | C9H11NO3 | 181.19 g/mol

An unnatural amino acid, that has demonstrated potential in treating Parkinsons disease, Alzheimers disease, and arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethyl sulfoxide, dilute acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerated (4°C).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 587-33-7
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00063059
InChI Key JZKXXXDKRQWDET-SVGMAFHSNA-N
Synonym l-m-tyrosine, 3-hydroxy-l-phenylalanine, meta-tyrosine, s-2-amino-3-3-hydroxyphenyl propanoic acid, 3-tyrosine, 3-hydroxyphenylalanine, m-tyrosine, l-m-tyr, unii-d5yf57v4qw, 3-m-hydroxyphenyl alanine
PubChem CID 6950578
ChEBI CHEBI:44303
IUPAC Name (2S)-2-amino-3-(3-hydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=CC(O)=C1)C(O)=O

Specifications

Melting Point 260°C to 270°C
Solubility Information Soluble in dimethyl sulfoxide,dilute acid.
Quantity 250 mg
Formula Weight 181.19
Percent Purity ≥97%
Chemical Name or Material L-m-Tyrosine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.