Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(-)-Malic acid, 99%

Catalog No. p-7042640
Click to view available options
Quantity:
100 g
500 g
2.5 kg

CAS: 97-67-6 | C4H6O5 | 134.087 g/mol

L-(-)-Malic acid is used as a food additive, Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid receptor agonists, 1α,25-dihydroxyvitamin D3 analogue, and phoslactomycin B.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(-)-Malic acid is used as a food additive, Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid receptor agonists, 1α,25-dihydroxyvitamin D3 analogue, and phoslactomycin B.

Solubility
Soluble in water(363g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-67-6
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.087
MDL Number MFCD00064213
InChI Key BJEPYKJPYRNKOW-REOHCLBHSA-N
Synonym l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid
PubChem CID 222656
ChEBI CHEBI:30797
IUPAC Name (2S)-2-hydroxybutanedioic acid
SMILES C(C(C(=O)O)O)C(=O)O

Specifications

Melting Point 100°C to 106°C
Color White
Density 1.6
Flash Point 220°C (428°F)
Quantity 100 g
Beilstein 1723541
Merck Index 14,5707
Solubility Information Soluble in water(363g/L).
Optical Rotation −26° (c=5.5 in Pyridine)
Formula Weight 134.09
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-Malic acid
Show More Show Less
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 202-601-5
RTECSNumber ON7175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.