Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich L-(-)-Malic Acid Sodium Salt

Catalog No. 503081501 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1501 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 50-308-1501 Supplier Sigma Aldrich Supplier No. M112525G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

L-Malic acid is the naturally occurring isomer of malic acid, found mainly in sour and unripe fruits.L-(-)-Malic acid sodium salt may be used to prepare standard solution during the analysis of wine samples using peroxidase based biosensors.

Specifications

CAS 68303-40-2
Melting Point 214°C
Molecular Formula C10H13NO2S
Linear Formula C6H5CH2SCH2CH(NH2)CO2H
MDL Number MFCD00002613
Quantity 10 g
Synonym L-Hydroxybutanedioic acid sodium salt
Molecular Weight (g/mol) 211.28 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.