Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(-)-Mannose, 99+%

Catalog No. AC227590010
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227590010 1 g Glass bottle
AC227592500 250 mg Glass bottle
AC227590050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC227590010 Supplier Thermo Scientific Chemicals Supplier No. 227590010
Add to Cart

CAS: 10030-80-5 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10030-80-5
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00136021
InChI Key GZCGUPFRVQAUEE-BXKVDMCESA-N
Synonym l-mannose, l---mannose, aldehydo-l-mannose, mannose, l, 2r,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, unii-2w3ye50tx8, aldehydo-l-manno-hexose, l-?-mannose
PubChem CID 82308
ChEBI CHEBI:37681
IUPAC Name (2R,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
SMILES OC[C@H](O)[C@H](O)[C@@H](O)[C@@H](O)C=O

Specifications

Melting Point 129.0°C to 131.0°C
Color White
Ignition Residue 0.1% max.
Loss on Drying 0.5% max. (60° to 70°C)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition −14.70° (20°C c=4,H2O,NH3)
Specific Rotation − 14.70
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol,insoluble in ether
Formula Weight 180.16
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material L(-)-Mannose
Show More Show Less
EINECSNumber 233-080-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.