Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Menthol, 99%

Catalog No. p-7080785
Click to view available options
Quantity:
50 g
250 g
1000 g

CAS: 2216-51-5 | C10H20O | 156.269 g/mol

L-Menthol is used as a cooling agent that strongly activates TRPM8.(Transient Receptor Potential Cation Channel, Subfamily M, Member 8 is a Protein Coding gene). It is used as analgesic (topical), antipruritic agent. It is used as: refreshing agent, food flavor, cool and antipruritic drug, carminative drug. Menthol crystals is used for pers onal care and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in 100% ethanol, methanol (100 mg/ml), ether, and chloroform. Insoluble in water.

Notes
Store at room temperature. Incompatible with Strong oxidizing agents, phenols, potassium permanganate, chromium trioxide, chloral hydrate, thymol, pyrogallol, resorcinol, camphor, butylchloral hydrate, betanaphthol, exalgine.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2216-51-5
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00062979
InChI Key NOOLISFMXDJSKH-KXUCPTDWSA-N
Synonym l-menthol, --menthol, levomenthol, menthomenthol, l---menthol, hexahydrothymol, menthacamphor, peppermint camphor, 1r,2s,5r-2-isopropyl-5-methylcyclohexanol, u.s.p. menthol
PubChem CID 16666
ChEBI CHEBI:15409
IUPAC Name (1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol
SMILES CC1CCC(C(C1)O)C(C)C

Specifications

Melting Point 42°C to 45°C
Density 0.89
Boiling Point 212°C to 216°C
Flash Point 93°C (199°F)
Quantity 50 g
Beilstein 1902293
Merck Index 14,5837
Solubility Information Soluble in 100% ethanol,methanol (100mg/ml),ether,and chloroform. Insoluble in water.
Optical Rotation −50° (c=10 in ethanol)
Formula Weight 156.27
Percent Purity 99%
Chemical Name or Material L-Menthol
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 218-690-9
RTECSNumber OT0700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.