Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Methionine, Multi-Compendial, U.S.P., J.T. Baker™

Catalog No. 02003230 Shop All Avantor Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-230 100 g Poly Bottle
02-003-231 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 02-003-230 Supplier Avantor Supplier No. 208505
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
PubChem CID 6137
ChEBI CHEBI:16643
SMILES CSCC[C@H](N)C(O)=O

Specifications

Packaging Poly Bottle
IUPAC Name (2S)-2-amino-4-(methylsulfanyl)butanoic acid
Quantity 100 g
Formula Weight 149.21
Grade Multi-Compendial/USP
Chemical Name or Material L-Methionine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.