Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Methionine tert-butyl ester hydrochloride, 95%, Thermo Scientific™

Catalog No. AAH65439MD
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
L-Methionine tert-butyl ester hydrochloride′s main function is to act as the primary “Start” sequence on mRNA so that the ribosomes can start translating the mRNA into proteins.

Notes
Store at 4°C. Incompatible with heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91183-71-0
Molecular Formula C9H20ClNO2S
Molecular Weight (g/mol) 241.77
MDL Number MFCD00038896
InChI Key KZJQROCWHZGZBJ-FJXQXJEOSA-N
Synonym h-met-otbu.hcl, h-met-otbu hcl, h-met-otbu hydrochloride, s-tert-butyl 2-amino-4-methylthio butanoate hydrochloride, l-methionine tert-butyl ester hydrochloride, tert-butyl l-methioninate-hydrogen chloride 1/1, l-methionine, 1,1-dimethylethyl ester, hydrochloride, 2-amino-4-methylsulfanyl-butyric acid tert-bu ester, hydrochloride, tert-butyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-met-otbu?hci
PubChem CID 13141795
IUPAC Name tert-butyl (2S)-2-amino-4-(methylsulfanyl)butanoate hydrochloride
SMILES Cl.CSCC[C@H](N)C(=O)OC(C)(C)C

Specifications

Beilstein 4884935
Quantity 250 mg
Formula Weight 241.77
Percent Purity 95%
Chemical Name or Material L-Methionine tert-butyl ester hydrochloride
TSCA No
Recommended Storage Keep cold
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.