Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ L-Myc Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566558
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566558 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566558 Supplier Invitrogen™ Supplier No. PA566558
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FELVPSPPTSP PWGLGPGAGD PAPGIGPPEP WPGGCTGDEA ESRGHSKGWG RNYAS Highest antigen sequence identity to the following orthologs - mouse 86%, rat 86%.

Cellular DNA-binding proteins encoded by the c-myc genes. They are normally involved in nucleic acid metabolism and in mediating the cellular response to growth factors. Elevated and deregulated (constitutive) expression of c-myc proteins can cause tumorigenesis. L-Myc gene ontology annotations include DNA binding function, DNA-binding transcription factor activity, RNA polymerase II-specific action, protein dimerization activity, and regulation of inner ear auditory receptor cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen L-Myc
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYCL
Gene Accession No. P12524
Gene Alias Avian myelocytomatosis viral (v-myc) related oncogene homolog 1, lung carcinoma derived (Lmyc); AW536278; bHLHe38; Class E basic helix-loop-helix protein 38; LMYC; L-MYC; Lmyc1; Lmyc-1; l-myc-1 proto-oncogene; MYCL; MYCL proto-oncogene, bHLH transcription factor; Mycl1; myc-related gene from lung cancer; Protein L-Myc; protein L-Myc-1; v-myc avian myelocytomatosis viral oncogene lung carcinoma derived homolog; V-myc myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog 1, lung carcinoma derived; v-myc myelocytomatosis viral oncogene homolog, lung carcinoma derived (avian)
Gene Symbols MYCL
Host Species Rabbit
Immunogen Recombinant Human L-Myc. Recombinant protein control fragment (Product #RP-105255).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4610
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.