Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ L-NAME hydrochloride
GREENER_CHOICESDP

Catalog No. 0665100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-651-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 06-651-00 Supplier Tocris Bioscience™ Supplier No. 0665/100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Non-selective NOS inhibitor

NO synthase inhibitor.

Chemical Identifiers

CAS 51298-62-5
Molecular Formula C7H16ClN5O4

Specifications

Quantity 100 mg
Formula Weight 269.69
Molecular Weight (g/mol) 269.69
Chemical Name or Material L-NAME hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.