Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-NIL hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
A selective inhibitor of inducible nitric oxide synthase (IC50 = 3.3 μM).
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 223.7 |
| Percent Purity | >98% |
| Chemical Name or Material | L-NIL hydrochloride |
Safety and Handling
| Recommended Storage | Desiccate at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction