Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ L-NMMA acetate
SDP

Catalog No. 077110 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-711-0 10 mg
07-715-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 07-711-0 Supplier R&D Systems™ Supplier No. 0771/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Non-selective NOS inhibitor

Competitive, irreversible inhibitor of all three NOS isoforms.

Chemical Identifiers

CAS 53308-83-1
Molecular Formula C9H20N4O4

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 248.28
Purity >98%
Molecular Weight (g/mol) 248.283
Chemical Name or Material L-NMMA acetate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.