Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Norleucine 99.0+%, TCI America™
SDP

Catalog No. N03031G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03031G 1 g
N0303100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N03031G Supplier TCI America Supplier No. N03031G

Chemical Identifiers

CAS 327-57-1
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064423
InChI Key LRQKBLKVPFOOQJ-YFKPBYRVSA-N
Synonym l-norleucine, norleucine, h-nle-oh, s-2-aminohexanoic acid, caprine, glycoleucine, l-+-norleucine, l-2-aminohexanoic acid, 2-aminocaproic acid, l-2-aminohexanoate
PubChem CID 21236
ChEBI CHEBI:18347
IUPAC Name (2S)-2-aminohexanoic acid
SMILES CCCC[C@H](N)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 131.18
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Norleucine
RTECSNumber RC6308000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.