Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Phenylalaninamide hydrochloride, 97%

Click to view available options
Quantity:
5g
25g
100g
65864-22-4
C9H13ClN2O
200.666
MFCD00039083
KLHLGTPNBQXSJT-QRPNPIFTSA-N
l-phenylalaninamide hydrochloride,h-phe-nh2.hcl,h-phe-nh2 hcl,l-phenylalanine amide hydrochloride,phenylalanine amide hydrochloride,h-phe-nh2hcl,2s-2-amino-3-phenylpropanamide hydrochloride,c9h12n2o.hcl,phenh2hcl,h-phe-nh hcl
11953719
(2S)-2-amino-3-phenylpropanamide;hydrochloride
C1=CC=C(C=C1)CC(C(=O)N)N.Cl
65864-22-4
C9H13ClN2O
200.666
MFCD00039083
KLHLGTPNBQXSJT-QRPNPIFTSA-N
l-phenylalaninamide hydrochloride,h-phe-nh2.hcl,h-phe-nh2 hcl,l-phenylalanine amide hydrochloride,phenylalanine amide hydrochloride,h-phe-nh2hcl,2s-2-amino-3-phenylpropanamide hydrochloride,c9h12n2o.hcl,phenh2hcl,h-phe-nh hcl
11953719
(2S)-2-amino-3-phenylpropanamide;hydrochloride
C1=CC=C(C=C1)CC(C(=O)N)N.Cl
Beilstein 3915662
Formula Weight 200.67
Physical Form Powder
Percent Purity 97%
Melting Point 231°C to 237°C
Quantity 25g
Chemical Name or Material L-Phenylalaninamide hydrochloride