Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Phenylpropionaldehyde, 98%

Catalog No. AC130865000
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 93-53-8 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-53-8
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00006973
InChI Key IQVAERDLDAZARL-UHFFFAOYSA-N
Synonym 2-phenylpropionaldehyde, hydratropaldehyde, hydratropic aldehyde, hyacinthal, cumene aldehyde, hydratropa aldehyde, hydrotropic aldehyde, 2-phenyl-1-propanal, 2-phenyl propionaldehyde, aldehyd hydratropovy
PubChem CID 7146
IUPAC Name 2-phenylpropanal
SMILES CC(C=O)C1=CC=CC=C1

Specifications

Melting Point -60.0°C
Color Colorless
Density 1.0110g/mL
Boiling Point 202.0°C to 205.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5160 to 1.5240
Linear Formula CH3CH(C6H5)CHO
Quantity 500 g
Beilstein 07, 305
Specific Gravity 1.011
Solubility Information Solubility in water: 3.4g/L (20°C).
Viscosity 2.16 mPa.s (20°C)
Formula Weight 134.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material DL-2-Phenylpropionaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of damaging fertility.
Causes serious eye irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protecti
EINECSNumber 202-255-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.