Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Phenylalanine dehydrogenase, MP Biomedicals™

Catalog No. MP219899410 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 U
50 U
75 U
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219899410 10 U
MP219899450 50 U
MP219899475 75 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219899410 Supplier MP Biomedicals, Inc Supplier No. 0219899410

L-Phenylalanine dehydrogenase >6 u/mg solid, Light Gray Powder

  • It is used to study phenylalanine metabolism and phenylalanine, tyrosine and tryptophan biosynthesis.
  • It is a NAD-dependent enzyme that catalyzes the reversible deamidation of L-phenylalanine into phenylpyruvate.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69403-12-9
Synonym E.C. 1.4.1.20

Specifications

Color Light Gray
Quantity 10 U
Physical Form Powder
storageNote1 Store at -20°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.