Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Plastin Monoclonal Antibody (LPL4A.1), Invitrogen™

Catalog No. PIMA511921
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA511921 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA511921 Supplier Thermo Scientific Supplier No. MA511921

Mouse Monoclonal Antibody

MA5-11921 targets Plastin-L in IF, IP, and WB applications and shows reactivity with Human samples. The MA5-11921 immunogen is human recombinant L-plastin protein.

L-Plastin Monoclonal antibody specifically detects L-Plastin in Human samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen L-Plastin
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone LPL4A.1
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene LCP1
Gene Accession No. P13796
Gene Alias 65 kDa macrophage protein; AW536232; bA139H14.1 (lymphocyte cytosolic protein 1 (L-plastin)); cb245; CP64; D14Ertd310e; DKFZp781A23186; epididymis secretory protein Li 37; fj24b12; FLJ25423; FLJ26114; FLJ39956; HEL-S-37; LC64P; LCP1; LCP-1; leucocyte-specific plastin 1; L-fimbrin; LPL; L-plastin; L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (65 kDa macrophage protein) (PP65); L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (LC64P); lymphocyte cytosolic plastin 1; lymphocyte cytosolic protein 1; lymphocyte cytosolic protein 1 (L-plastin); Lymphocyte cytosolic protein-1 (plasmin); plastin 2; plastin 2, L; plastin-2; Pls2; pp65; wu:fj24b12
Gene Symbols LCP1
Host Species Mouse
Immunogen Human recombinant L-plastin protein
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3936
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.