Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ L-Plastin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597918
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597918 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597918 Supplier Invitrogen™ Supplier No. PA597918
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

L-Plastin Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA

L-plastin is a leukocyte specific actin bundling protein from fimbrin family of proteins. It has two actin-binding domains and two EF hand type calcium-binding domains. It gets phosphorylated on the serine residue in its headpiece region in response to a variety of inflammatory stimuli. It is hypothesized that L-plastin may play a role in regulating integrin mediated adhesion in leukocytes. L-plastin is expressed in majority of human cancer cell lines derived from solid tumors. Its expression is positively upregulated by testosterones in androgen receptor positive prostrate and breast cancer cells through three androgen receptor binding elements located in the upstream region of the gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen L-Plastin
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 2.665 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LCP1
Gene Accession No. P13796
Gene Alias 65 kDa macrophage protein; AW536232; bA139H14.1 (lymphocyte cytosolic protein 1 (L-plastin)); cb245; CP64; D14Ertd310e; DKFZp781A23186; epididymis secretory protein Li 37; fj24b12; FLJ25423; FLJ26114; FLJ39956; HEL-S-37; LC64P; LCP1; LCP-1; leucocyte-specific plastin 1; L-fimbrin; LPL; L-plastin; L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (65 kDa macrophage protein) (PP65); L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (LC64P); lymphocyte cytosolic plastin 1; lymphocyte cytosolic protein 1; lymphocyte cytosolic protein 1 (L-plastin); Lymphocyte cytosolic protein-1 (plasmin); plastin 2; plastin 2, L; plastin-2; Pls2; pp65; wu:fj24b12
Gene Symbols LCP1
Host Species Rabbit
Immunogen Recombinant Human Plastin-2 protein (2-627AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3936
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.