Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Prolinanilide 98.0+%, TCI America™
SDP

Catalog No. P24301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24301G 1 g
P24305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P24301G Supplier TCI America Supplier No. P24301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64030-43-9
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.246
MDL Number MFCD08276245
InChI Key NRXYBKZDRCXDOD-JTQLQIEISA-N
Synonym (S)-N-Phenylpyrrolidine-2-carboxamide
PubChem CID 6552381
IUPAC Name (2S)-N-phenylpyrrolidine-2-carboxamide
SMILES C1CC(NC1)C(=O)NC2=CC=CC=C2

Specifications

Melting Point 78°C
Color White-Yellow
Quantity 1 g
Formula Weight 190.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Prolinanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.