Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Serine, 98.5%, Spectrum™ Chemical

Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Poly Pail
Specifications
Specifications
CAS Max % | 1 |
Lead (Pb) | 10 ppm |
Loss on Drying | 0.003 |
Packaging | Poly Pail |
Specific Rotation | +13.6° to +15.6° |
Quantity | 1 kg |
Percent Purity | 98.5% |
Grade | Reagent |
Chemical Name or Material | L-Serine |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction