Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Proline Ethyl Ester, TCI America™
SDP

Catalog No. P12471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12471G 1 g
P12475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12471G Supplier TCI America Supplier No. P12471G

Chemical Identifiers

CAS 5817-26-5
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD11656706
InChI Key QPNJHVDIRZNKOX-LURJTMIESA-N
Synonym H-Pro-OEt
PubChem CID 79888
IUPAC Name ethyl (2S)-pyrrolidine-2-carboxylate
SMILES CCOC(=O)[C@@H]1CCCN1

Specifications

Color Yellow
Boiling Point 83°C
Quantity 1 g
Formula Weight 143.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material L-Proline Ethyl Ester
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.